Learn More
Tocris Bioscience™ RU-SKI 43 hydrochloride
Hedgehog acyltransferase (Hhat) inhibitor; cell permeable
Description
Description
Hedgehog acyltransferase (Hhat) inhibitor (IC50 = 850 nM). Inhibits Sonic Hedgehog N (ShhN) palmitoylation in vitro. Has no effect on porcupine-induced fatty acid acylation of Wnt39a. Attenuates pancreatic cancer cell proliferation. Cell permeable.