missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ RU.521

Description
RU.521 is a potent and selective cyclic GMP-AMP synthase (cGAS) inhibitor (Kd = 36.2 nM; IC50 values are 0.11 μM and 2.94 μM against m-cGAS and h-cGAS, respectively). RU.521 suppresses dsDNA-activated reporter activity but not 5'ppp-HP20 RNA or murine IFN-β-induced IFN-β1-dependent gene expression. RU.521 reduces constitutive expression of type I interferon in primary macrophages from a mouse model of Aicardi-Gouti?res syndrome. Pretreatment of chimeric antigen receptor (CAR) redirected T cells with RU.521 increases efficiency of virus-free gene editing by improving cell viability and CAR integration in cell line.
Specifications
Specifications
| Chemical Name or Material | 3-[1-(6,7-Dichloro-1 H-benzimidazol-2-yl)-5-hydroxy-3-methyl-1 H-pyrazol-4-yl]-1(3 H)-isobenzofuranone |
| CAS | 2262452-06-0 |
| Quantity | 50 mg |
| Target | Synthase/Synthetase Inhibitors |
| Molecular Formula | C19H12Cl2N4O3 |
| Purity | 0.98 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?