missing translation for 'onlineSavingsMsg'
Learn More

RU 24969, MedChemExpress

Product Code. 16493157
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16493157 50 mg 50mg
16487128 10 mg 10mg
16469667 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16493157 Supplier Medchem Express Supplier No. HY16688

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress RU 24969 is a preferential 5-HT1B agonist, with a Ki of 0.38 nM, but also displays appreciable affinity for the 5-HT1A receptor (Ki=2.5 nM), and has low affinity for other receptor sites in the brain. RU 24969 could decrease fluid consumption and increase forward locomotion.

RU 24969 is a preferential 5-HT1B agonist, with a Ki of 0.38 nM, but also displays appreciable affinity for the 5-HT1A receptor (Ki=2.5 nM), and has low affinity for other receptor sites in the brain. RU 24969 could decrease fluid consumption and increase forward locomotion.

Specifications

Chemical Name or Material RU 24969
CAS 66611-26-5
Color Yellow
Purity Grade Notes Research
Molecular Formula C14H16N2O
Quantity 50 mg
Solubility Information DMSO : ≥ 30 mg/mL (131.41 mM)
SMILES COC1=CC=C2C(C(C3=CCNCC3)=CN2)=C1
Molecular Weight (g/mol) 228.29
Formula Weight 228.29
Percent Purity 99.77%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.