missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ RU 24969 hemisuccinate
GREENER_CHOICE

Product Code. 18769966
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18769966

Brand: Tocris Bioscience™ 0912/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

5-HT1A and 5-HT1B agonist

RU 24969 hemisuccinate is a potent 5-HT1A/1B and moderate 5-HT2C agonist that may also release 5-HT. Centrally active following systemic administration.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Methoxy-3-(1,2,5,6-tetrahydro-4-pyridinyl)-1H-indole hemisuccinate
CAS 66611-27-6
Assay Percent Range >99%
Quantity 10 mg
Solubility Soluble to 50mM in DMSO
Molecular Formula C14H16N2O.½C4H6O4
Formula Weight Observed MW: 287.34

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.