missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ RU 24969 hemisuccinate

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
RU 24969 hemisuccinate is a potent 5-HT1A/1B and moderate 5-HT2C agonist that may also release 5-HT. Centrally active following systemic administration.
Specifications
Specifications
| Chemical Name or Material | 5-Methoxy-3-(1,2,5,6-tetrahydro-4-pyridinyl)-1H-indole hemisuccinate |
| CAS | 66611-27-6 |
| Assay Percent Range | >99% |
| Quantity | 50 mg |
| Solubility | Soluble to 50mM in DMSO |
| Molecular Formula | C14H16N2O.½C4H6O4 |
| Formula Weight | Observed MW: 287.34 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction