missing translation for 'onlineSavingsMsg'
Learn More

Restek™ Rtx-20 GC Capillary Columns

Product Code. 13011150
Encompass_Preferred
Click to view available options
Film Thickness:
1 μm
Particle Size:
1 μm
This item is not returnable. View return policy

Product Code. 13011150

Brand: Restek™ 10355

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Rtx-20 polymer is synthesized to exacting standards. All residual catalysts and low molecular weight fragments are removed from the polymer, providing a tight monomodal distribution and extremely low bleed.

  • General-purpose columns for volatile compounds, flavor compounds, alcoholic beverages
  • Equivalent to USP G28, G32 phases.

Specifications

Max. Temperature 350°C
Type Capillary Column
Diameter (Metric) Inner 0.53 mm
Stationary Phase Rtx™-20
USP Type G28, G32
For Use With (Application) Volatile Compounds Analysis, Flavor Compounds Analysis, Alcoholic Beverages Analysis
Particle Size 1 μm
Temperature Range -20°C to +260/280°C
Film Thickness 1 μm
Length (Metric) 30 m
Material Stainless Steel
Polarity Low to mid polar
Coating Fused Silica
Quantity 1 Each
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.