missing translation for 'onlineSavingsMsg'
Learn More

Diversey™ RTU Powergel VG11 Cleaner

Product Code. 17683245
Click to view available options
Volume (Metric):
20 L
Unit Size:
20 L
This item is not returnable. View return policy

Product Code. 17683245

Brand: Diversey™ 7522708

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ready-to-use heavy duty alkaline cleaner with sequestrants for polymerized soil.

RTU Powergel is a caustic-based cleaner containing sequestrants and surfactants/ wetting agents. This blend has been shown to be particularly effective for removing polymerized oil from spiral and tunnel ovens, batch cookers, fryers and other area’s with polymerised and/or carbonised soil.

Benefits

  • RTU Powergel enables an easy cleaning of difficult soils like polymerized oil which has turned into high viscous or solid soiling.
  • Application of this product on surfaces may lead to a decrease of chemical consumption in later stages of cleaning.
  • Reduction of the cleaning time and manual cleaning.

Specifications

Form Gel
pH >13
Sterility Non-sterile
Composition Caustic-based cleaner containing sequestrants and surfactants/ wetting agents
Container Type Canister
Volume (Metric) 20 L
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.