missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RTP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18973-1-AP-20UL
This item is not returnable.
View return policy
Description
Specifically promotes functional cell surface expression of olfactory receptors, but not of other GPCRs.Specifications
| RTP1 | |
| Polyclonal | |
| Unconjugated | |
| RTP1 | |
| RTP1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 132112, 239766, 288006 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P59025, Q8C8C1 | |
| Rtp1 | |
| RTP1 Fusion Protein Ag6934 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction