missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RTN4RL2 Monoclonal Antibody (333413)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561203
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
Reticulon 4 receptor-like 2 (RTN4RL2), also known as NgR2and NgRH1, is a 60 kDa glycosylphosphatidylinositol-anchored protein that is expressed primarily in the brain. It contains eight leucine-rich repeats that are flanked by cysteine-rich sequences at both the N- and C-termini. The membrane anchored RTN4RL2 can be solubilized through the action of phospholipase or an unidentified MTMMP to generate a 46 kDa soluble receptor. It has been suggested that RTN4RL2 may play a role in regulating axonal regeneration and plasticity in the adult central nervous system.
Specifications
| RTN4RL2 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q86UN3 | |
| RTN4RL2 | |
| Mouse myeloma cell line NS0-derived recombinant human NgR2/NgRH1, Ala28-Leu401. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Western Blot | |
| 333413 | |
| Unconjugated | |
| RTN4RL2 | |
| ngR2; NGRH1; NGRL3; Nogo receptor-like 3; Nogo-66 receptor homolog 1; Nogo-66 receptor-related protein 2; reticulon 4 receptor-like 2; reticulon-4 receptor-like 2; RGD:727797}; Rtn4rl2; rtn4rl2 {ECO:0000312 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 349667 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction