missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RTN4IP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557625
This item is not returnable.
View return policy
Description
Immunogen sequence: TLVTPFLLNM DRLGIADGML QTGVTVGSKA LKHFWKGVHY RWAFFMASGP CLDDIAELVD AGKIRPVIEQ TFPFSKVPEA FLKVERGHAR GK Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 91%.
This gene encodes a novel mitochondrial protein that interacts with reticulon 4, which is a potent inhibitor of regeneration following spinal cord injury. The interaction of reticulon 4 with mitochondrial proteins may provide insight into the mechanisms for reticulon-induced inhibition of neurite growth.
Specifications
| RTN4IP1 | |
| Polyclonal | |
| Unconjugated | |
| Rtn4ip1 | |
| D10Ertd690e; Nimp; NOGO-interacting mitochondrial protein; OPA10; reticulon 4 interacting protein 1; reticulon 4 interacting protein 1, mitochondrial; reticulon-4-interacting protein 1, mitochondrial; RGD1563384; RTN4IP1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84816 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8WWV3 | |
| Rtn4ip1 | |
| Recombinant protein corresponding to Human RTN4IP1. Recombinant protein control fragment (Product #RP-94819). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction