missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RTN4IP1 Antibody (OTI3B2), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73952MFV500
This item is not returnable.
View return policy
Description
RTN4IP1 Monoclonal antibody specifically detects RTN4IP1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry
Specifications
| RTN4IP1 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Flow Cytometry | |
| OTI3B2 | |
| Flow Cytometry | |
| MGC12934, NIMPNOGO-interacting mitochondrial protein, reticulon 4 interacting protein 1, reticulon-4-interacting protein 1, mitochondrial | |
| Full length human recombinant protein of human RTN4IP1 (NP_116119) produced in E.coli. | |
| 0.1 mL | |
| metabolism | |
| 84816 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction