missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RTN4/NOGO Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10950-1-AP-150UL
This item is not returnable.
View return policy
Description
NogoA is a member of a family of integral membrane proteins termed reticulons that are thought to be involved in numerous disorders including neurodegenerative diseases. Reticulon proteins are known to regulate many cellular processes and interact with multiple proteins and receptors such as BACE. NogoA was initially identified as a myelin-associated neurite outgrowth inhibitor. It is highly expressed in oligodendrocytes in the white matter of the CNS, blocking its activity with antibodies or other factors results in improved axon regrowth and functional recovery in experimental CNS lesion models. NogoA has also been suggested to play a role in neurodegenerative diseases such as Amyotrophic lateral sclerosis, in which case NogoA is found at elevated levels in postmortem muscular samples, and multiple sclerosis (MS), in which case autoantibodies to NogoA have been found in serum and cerebrospinal fluid in MS patients.Specifications
| RTN4/NOGO | |
| Polyclonal | |
| Unconjugated | |
| RTN4 | |
| ASY, Foocen, KIAA0886, Nbla00271, Nbla10545, Neurite outgrowth inhibitor, NI220/250, NOGO, NOGO A, Nogo B, Nogo C, Nogo protein, NOGOA, NOGOC, NSP, NSP CL, reticulon 4, Reticulon 5, RTN X, RTN4, RTN4 A, RTN4 B1, RTN4 B2, RTN4 C, SP1507 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 57142 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9NQC3 | |
| RTN4 | |
| RTN4/NOGO Fusion Protein Ag1392 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction