missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RTN3 Monoclonal Antibody (10A8)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA515528
This item is not returnable.
View return policy
Description
MA5-15528 targets RTN3 in indirect ELISA, IHC applications and shows reactivity with Human samples. The MA5-15528 immunogen is purified recombinant fragment of RTN3 expressed in E. Coli. MA5-15528 detects RTN3 which has a predicted molecular weight of approximately 113kDa.
RTN3 is a member of reticulons, which are associated with the endoplasmic reticulum, and are involved in neuroendocrine secretion or in membrane trafficking in neuroendocrine cells. Alternatively spliced transcript variants encoding different isoforms have been identified. Multiple promoters rather than alternative splicing of internal exons seem to be involved in this diversity.
Specifications
| RTN3 | |
| Monoclonal | |
| Conc. Not Determined | |
| Ascites with 0.03% sodium azide | |
| O95197 | |
| RTN3 | |
| Purified recombinant fragment of RTN3 expressed in E. Coli. | |
| RUO | |
| 10313 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin) | |
| 10A8 | |
| Unconjugated | |
| RTN3 | |
| ASY interacting protein; ASYIP; HAP; Homolog of ASY protein; neuroendocrine-specific protein-like 2; Neuroendocrine-specific protein-like II; nsp-c; NSPL2; NSPLII; Nsp-like II; NSP-like protein 2; NSP-like protein II; reticulon 3; reticulon-3; RTN3; RTN3-A1 | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto