missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RTN2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5107018
This item is not returnable.
View return policy
Description
Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 93%; Rat: 100%.
This gene belongs to the family of reticulon encoding genes. Reticulons are associated with the endoplasmic reticulum, and are involved in neuroendocrine secretion or in membrane trafficking in neuroendocrine cells. Alternatively spliced transcript variants encoding different isoforms have been identified.
Specifications
| RTN2 | |
| Polyclonal | |
| Unconjugated | |
| RTN2 | |
| formerly RTN2w; MMS10-P; Ms10p; neuroendocrine-specific protein-like 1; Neuroendocrine-specific protein-like I; NSP2; Nspl1; NSPLI; NSP-like 1; NSP-like protein 1; NSP-like protein I; reticulon 2; reticulon 2 (Z-band associated protein); reticulon-2; RTN2; RTN2-B; SPG12 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 6253 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 | |
| O75298 | |
| RTN2 | |
| Synthetic peptide directed towards the N terminal region of human RTN2 (aa 1-51). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction