missing translation for 'onlineSavingsMsg'
Learn More

RTN1-A/NSP Antibody (MON162), Biotin, Novus Biologicals™

Product Code. 18405701 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18405701 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18405701 Supplier Novus Biologicals Supplier No. NBP197677B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

RTN1-A/NSP Monoclonal antibody specifically detects RTN1-A/NSP in Human, Mouse, Rat, Hamster samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
TRUSTED_SUSTAINABILITY

Specifications

Antigen RTN1-A/NSP
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone MON162
Conjugate Biotin
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation PBS
Gene Alias NSP
Host Species Mouse
Immunogen Derived by fusion of Mouse myeloma cells with spleen cells from a Mouse immunized with a partially purified bacterially expressed Reticulon-1A (NSP-A) hybrid protein.
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 6252
Target Species Human, Mouse, Rat, Hamster
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.