missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ RTKN2 293T Cell Transient Overexpression Lysate

Product Code. 18407689 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18407689 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18407689 Supplier Novus Biologicals™ Supplier No. H00219790T01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Quantity 0.1 mL
Content And Storage Store at -80°C. Avoid freeze/thaw cycles.
Target RTKN2
Synonym bA531F24.1, DKFZp686J10120, Em:AC024597.2, FLJ39352, PH domain-containing family K member 1, pleckstrin homology domain containing, family K member 1, Pleckstrin homology domain-containing family K member 1, PLEKHK1, rhotekin 2, rhotekin-2
Gene Symbol RTKN2
Product Title
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.