missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RTI-13951-33 hydrochloride, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Description
RTI-13951-33 hydrochloride is a potent, selective, and brain-penetrant GPR88 agonist, with an EC50 of 25 nM in GPR88 cAMP functional assay. RTI-13951-33 hydrochloride reduces alcohol reinforcement and intake behaviors in rats.
Specifications
Specifications
| Chemical Name or Material | RTI-13951-33 hydrochloride |
| Physical Form | Solid |
| Color | Light Yellow |
| Quantity | 50 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C28H35Cl2N3O3 |
| For Use With (Application) | Neuroscience-Neuromodulation |
| Solubility Information | DMSO : 50 mg/mL (93.90 mM; Need ultrasonic) ∣H2O : 50 mg/mL (93.90 mM; Need ultrasonic) |
| SMILES | COCC1=CC=C(C2=CC=C(N(C[C@@H](N)[C@@H](C)OC)C([C@H]3[C@H](C4=CC=CC=N4)C3)=O)C=C2)C=C1.Cl.Cl |
| Molecular Weight (g/mol) | 532.5 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction