missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RTF1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552330
This item is not returnable.
View return policy
Description
Immunogen sequence: GNHNSKPVYR VAEITGVVET AKVYQLGGTR TNKGLQLRHG NDQRVFRLEF VSNQEFTESE FMKWKEAMFS AGMQLPTLDE INKKELSIKE ALNYKFNDQD IEEIVKEKER FRKAPPNYAM KKTQL Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 99%.
Three isoforms of serine/threonine kinases, designated alphaPAK p68, betaPAK p65 and gammaPAK p62, have been shown to exhibit a high degree of sequence homology with the S. cerevisiae kinase Ste 20, involved in pheromone signaling. The alpha, beta and gammaPAK isoforms complex specifically with Rac1 and Cdc42 in their active GTP-bound state, inhibiting their intrinsic GTPase activity leading to their autophosphorylation. There are eight sites of autophosphorylation on gammaPAK, including Ser 19, Ser 141 and Thr 402, and phosphorylation of Ser 141 and Thr 402 is correlated with gammaPAK activation. Once phosphorylated and their affinity for Rac/Cdc42 reduced, the PAK isoforms disassociate from the complex to seek downstream substrates.
Specifications
| RTF1 | |
| Polyclonal | |
| Unconjugated | |
| RTF1 | |
| 2900005O08Rik; 6530416A09Rik; AI853581; AU043144; AW553985; cbp82; CG10955; CG10955-PA; CG10955-PB; Dmel\CG10955; Dmel_CG10955; dRtf1; gene trap locus 7; Gtl7; KIAA0252; ortholog of mouse gene trap locus 7; RNA polymerase-associated protein Rtf1; RNA polymerase-associated protein RTF1 homolog; Rtf1; RTF1 homolog, Paf1/RNA polymerase II complex component; Rtf1 Paf1/RNA polymerase II complex component homolog; Rtf1, Paf1/RNA polymerase II complex component, homolog; Rtf1, Paf1/RNA polymerase II complex component, homolog (S. cerevisiae); Rtf1-PA; Rtf1-PB | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23168 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q92541 | |
| RTF1 | |
| Recombinant protein corresponding to Human RTF1. Recombinant protein control fragment (Product #RP-89460). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction