missing translation for 'onlineSavingsMsg'
Learn More

RTF1 Antibody, Alexa Fluor™ 750, Novus Biologicals™

Product Code. 18421433 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18421433 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18421433 Supplier Novus Biologicals Supplier No. NB600277AF750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RTF1 Polyclonal antibody specifically detects RTF1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen RTF1
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 750
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias KIAA0252GTL7, ortholog of mouse gene trap locus 7, RNA polymerase-associated protein RTF1 homolog, Rtf1, Paf1/RNA polymerase II complex component, homolog (S. cerevisiae)
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residues 300 and 350 of human Restores TBP (TATA box-binding protein) function-1 using the numbering given in entry EAW92492.1 (GeneID 23168).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 23168
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.