missing translation for 'onlineSavingsMsg'
Learn More

RTEL1 Antibody, mFluor Violet 500 SE, Novus Biologicals™

Product Code. 18415237
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18415237 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18415237 Supplier Novus Biologicals (Bio-Techne) Supplier No. NBP222360MFV500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RTEL1 Polyclonal antibody specifically detects RTEL1 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen RTEL1
Applications Western Blot, Immunocytochemistry, Immunoprecipitation
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Formulation 50mM Sodium Borate
Gene Alias C20orf41, regulator of telomere elongation helicase 1, regulator of telomere length
Host Species Rabbit
Immunogen A genomic peptide made to an internal region of the human RTEL1 protein (within residues 800-1000). [Swiss-Prot Q9NZ71]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 51750
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.