missing translation for 'onlineSavingsMsg'
Learn More

RTCD1 Antibody, Novus Biologicals™

Product Code. 18358729 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18358729 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18358729 Supplier Novus Biologicals Supplier No. H00008634B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

RTCD1 Polyclonal antibody specifically detects RTCD1 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen RTCD1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS (pH 7.4)
Gene Accession No. NP_003720
Gene Alias phosphate cyclase, RNA 3'-terminal phosphate cyclase, RNA cyclase, RNA terminal phosphate cyclase domain 1, RNA terminal phosphate cyclase domain-containing protein 1, RNA-3'-phosphate cyclase, RPC1, RPCEC 6.5.1.4, RTC domain containing 1, RTC domain-containing protein 1, RTC1
Host Species Mouse
Immunogen RTCD1 (NP_003720, 1 a.a. - 366 a.a.) full-length human protein. MAGPRVEVDGSIMEGGGQILRVSTALSCLLGLPLRVQKIRAGRSTPGLRPQHLSGLEMIRDLCDGQLEGAEIGSTEITFTPEKIKGGIHTADTKTAGSVCLLMQVSMPCVLFAASPSELHLKGGTNAEMAPQIDYTVMVFKPIVEKFGFIFNCDIKTRGYYPKGGGEVIVRMSPVKQLNPINLTERGCVTKIYGRAFVAGVLPFKVAKDMAAAAVRCIRKEIRDLYVNIQPVQEPKDQAFGNGNGIIIIAETSTGCLFAGSSLGKRGVNADKVGIEAAEMLLANLRHGGTVDEYLQDQLIVFMALANGVSRIKTGPVTLHTQTAIHFAEQIAKAKFIVKKSEDEEDAAKDTYIIECQGIGMTNPNL
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 8634
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.