missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RtcB Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19809-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody specifically recognizes endogenous RTCB protein.
The function of this protein remains unknown.Specifications
| RtcB | |
| Polyclonal | |
| Unconjugated | |
| RTCB | |
| C22orf28, FAAP, HSPC117, RtcB | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 28088, 51493 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Western Blot, Immunoprecipitation, Immunofluorescence | |
| 0.18 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q99LF4, Q9Y3I0 | |
| Rtcb | |
| RtcB-Specific Fusion Protein Ag13836 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction