missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RTBDN Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA590530
This item is not returnable.
View return policy
Description
Immunogen sequence: SRPLQARSQQ HHGLAADLGK GKLHLAGPCC PSEMDTTETS GPGNHPERCG VPSPECESFL EHLQRALRSR FRLRLLGVRQ AQPLCEELCQ AWFANCEDDI TCGPTWLPLS EKRGCEPSCL TYGQTFADGT DLCRSALGHA LPVAAPGARH CFNISISAVP RPRPGRRGRE APSRRSRSPR TSILDAAGSG SGSGSGSGP; Positive Samples: Rat eye.
This gene was first identified in a study of human eye tissues. The protein encoded by this gene is preferentially expressed in the retina and may play a role in binding retinoids and other carotenoids as it shares homology with riboflavin binding proteins. Alternative splicing results in multiple transcript variants and protein isoforms. [provided by RefSeq, Jul 2012].
Specifications
| RTBDN | |
| Polyclonal | |
| Unconjugated | |
| RTBDN | |
| FLJ36353; retbindin; RTBDN; Rtbnd | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 304667, 83546 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 3.19 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q9BSG5 | |
| RTBDN | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 63-261 of human RTBDN (NP_1136171). | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction