missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RTBDN Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93185-0.02ml
This item is not returnable.
View return policy
Description
RTBDN Polyclonal antibody specifically detects RTBDN in Rat samples. It is validated for Western Blot
Specifications
| RTBDN | |
| Polyclonal | |
| Western Blot 1:200-1:2000 | |
| FLJ36353, retbindin | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 63-261 of human RTBDN (NP_113617.1). SRPLQARSQQHHGLAADLGKGKLHLAGPCCPSEMDTTETSGPGNHPERCGVPSPECESFLEHLQRALRSRFRLRLLGVRQAQPLCEELCQAWFANCEDDITCGPTWLPLSEKRGCEPSCLTYGQTFADGTDLCRSALGHALPVAAPGARHCFNISISAVPRPRPGRRGREAPSRRSRSPRTSILDAAGSGSGSGSGSGP | |
| 0.02 mL | |
| Neuroscience | |
| 83546 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction