Learn More
Invitrogen™ RT1.Bu Monoclonal Antibody (OX-3), PE
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA517419
Description
anti-rat RT1.Bu monoclonal antibody recognizes a polymorphic determinant on rat Ia present on Lewis, Wistar, and AO rat strains (although it is apparently expressed in lower amounts in AO rats) but not BN, DA, or PVG/c strains. This antibody also cross-reacts with mouse strains (MHC haplotypes b and s) and analysis of recombinant mouse strains shows that the determinant maps to the I-A region and correlates with mouse Ia specificity 9. Recognizes Ia antigens on approximately 18% of thymocytes but does not significantly label the majority of peripheral T lymphocytes. Thus, this antibody is particularly useful for distinguishing Ia positive cells from different rat strains and has been used in the recognition of cells of donor origin in bone marrow reconstituted radiation chimeras.
RT1.Bu is a gene located within the major histocompatibility complex (MHC) region of the rat genome, specifically part of the RT1 complex. The RT1 complex in rats is analogous to the human HLA and mouse H2 MHC regions and encompasses a variety of class I and class II genes involved in antigen presentation. RT1.Bu is one such class I MHC gene, encoding a protein essential for the presentation of endogenous peptides to cytotoxic T cells. This interaction is vital for the immune system's ability to recognize and eliminate infected or malignantly transformed cells. The RT1.Bu gene is crucial for studying immune responses, particularly in transplantation immunology and autoimmune disease models, due to its role in histocompatibility and immune regulation.
Specifications
| RT1.Bu | |
| Monoclonal | |
| 0.1 mg/mL | |
| PBS with 4-5mg/mL BSA and 0.02% sodium azide | |
| P20037 | |
| RT1-Ba | |
| Rat thymocyte membrane glycoprotein. | |
| 200 μg | |
| Primary | |
| Rat | |
| Antibody | |
| IgG1 |
| Flow Cytometry | |
| OX-3 | |
| PE | |
| RT1-Ba | |
| activated B cell RT1Bl alpha chain; BA1; MHC class II antigen RT1.B alpha chain; rano class II histocompatibility antigen, B alpha chain; rat class I Bu; RT1 class II antigen, Ba chain; RT1 class II, locus Ba; RT1-B alpha; RT1-Ba; RT1-Bu | |
| Mouse | |
| Protein G | |
| RUO | |
| 309621 | |
| 4°C, do not freeze | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.