missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RSV Polyclonal Antibody, Biotin

Description
The PA1-73019 antibody reacts with Respiratory Syncytial virus. PA1-73019 has been successfully used in Immunohistochemical, ELISA, Immunofluorescence and Western blot applications. The PA1-73019 antibody was raised against Human RSV isolate (confirmed).
Human respiratory syncytial virus (RSV) is a member of the Paramyxoviridae family, subfamily Pneumovirinae, that causes common respiratory tract infections such as measles and mumps. Its genome consists of single stranded, negative sense RNA that encodes three envelope glycoproteins, a small hydrophobic (SH) protein of unknown function, a major glycoprotein (G) known as the attachment protein, and a fusion (F) protein. Its name comes from the fact that F proteins on the surface of the virus cause the membranes on nearby cells to merge, forming syncytia. While RSV typically produces mild symptoms in adults, it is one of the leading causes of lower respiratory tract infections and hospital visits during infancy and childhood.
Specifications
Specifications
| Antigen | RSV |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 5 mg/mL |
| Conjugate | Biotin |
| Formulation | PBS with 0.1% sodium azide; pH 7.2 |
| Gene Alias | human respiratory syncytial virus; RSV |
| Host Species | Goat |
| Immunogen | Human RSV isolate (confirmed). |
| Purification Method | IgG fraction |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?