missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RSV NP Recombinant Rabbit Monoclonal Antibody (HL1295)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556857
This item is not returnable.
View return policy
Description
RSV NP Recombinant Monoclonal Antibody for Western Blot, ICC/IF, ELISA
Specifications
| RSV NP | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Rabbit | |
| Protein A | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
| ELISA, Western Blot, Immunocytochemistry | |
| HL1295 | |
| Unconjugated | |
| human respiratory syncytial virus; RSV; RSV NP; RSV Nuclear Protein | |
| Recombinant protein encompassing a sequence within the center region of Respiratory Syncytial virus Nucleoprotein. (Strain A2). | |
| 100 μL | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction