missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RSV M2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 11540893
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11540893 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11540893 Supplier Invitrogen™ Supplier No. PA123415

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PA1-23415 detects Respiratory Syncytial Virus M2 Protein in Virus samples.

The RSV (Respiratory Syncytial Virus) M2 gene encodes for two critical proteins, M2-1 and M2-2. The M2-1 protein is essential for viral transcription, acting as an elongation factor, ensuring the synthesis of full-length viral mRNAs. It associates non-covalently with the viral RNA-dependent RNA polymerase complex. Transcription regulation is influenced by the phosphorylation state of M2-1, mediated by cellular protein phosphatase 1 (PP1). In contrast, the M2-2 protein plays a significant role in regulating the balance between viral transcription and replication. These functions underscore the crucial roles of M2 gene products in the RSV lifecycle and their importance as potential targets for antiviral strategies.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RSV M2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Alias human respiratory syncytial virus; RSV; RSV M2
Host Species Rabbit
Immunogen Human Respiratory Syncytial Virus (RSV) M2 protein.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.