missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RSV Fusion Protein Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA522824

Product Code. 13287988

  • £480.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

RSV Fusion Protein Polyclonal Antibody for Western Blot, Flow

Human respiratory syncytial virus (RSV) is a member of the Paramyxoviridae family, subfamily Pneumovirinae, that causes common respiratory tract infections such as measles and mumps. Its genome consists of single stranded, negative sense RNA that encodes three envelope glycoproteins, a small hydrophobic (SH) protein of unknown function, a major glycoprotein (G) known as the attachment protein, and a fusion (F) protein. The 63 kD RSV fusion protein of the RSS 2 strain (subtype A) directs fusion of viral and cellular membranes, resulting in viral penetration, and can direct fusion of infected cells with adjoining cells, resulting in the formation of syncytia or multi nucleated giant cells. Its name comes from the fact that F proteins on the surface of the virus cause the membranes on nearby cells to merge, forming syncytia. While RSV typically produces mild symptoms in adults, it is one of the leading causes of lower respiratory tract infections and hospital visits during infancy and childhood.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RSV Fusion Protein
Polyclonal
Unconjugated
human respiratory syncytial virus; RSV; RSV Fusion protein; Rsvgp08
Residues 201-211 of the 63 kDa BRSV F protein.
100 μg
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Flow Cytometry, Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Rabbit
Antigen affinity chromatography
RUO
Virus
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.