Learn More
Invitrogen™ RSV Fusion Protein Monoclonal Antibody (D74G)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA17290
Description
The MA1-7290 antibody reacts with Respiratory Sync. virus fusion protein in viral and infected tissue samples. MA1-7290 has been successfully used in ELISA,immunofluorescence and inhibition assay applications. The MA1-7290 antibody was raised against an RSV fusion protein.
Human respiratory syncytial virus (RSV) is a member of the Paramyxoviridae family, subfamily Pneumovirinae, that causes common respiratory tract infections such as measles and mumps. Its genome consists of single stranded, negative sense RNA that encodes three envelope glycoproteins, a small hydrophobic (SH) protein of unknown function, a major glycoprotein (G) known as the attachment protein, and a fusion (F) protein. The 63 kD RSV fusion protein of the RSS 2 strain (subtype A) directs fusion of viral and cellular membranes, resulting in viral penetration, and can direct fusion of infected cells with adjoining cells, resulting in the formation of syncytia or multi nucleated giant cells. Its name comes from the fact that F proteins on the surface of the virus cause the membranes on nearby cells to merge, forming syncytia. While RSV typically produces mild symptoms in adults, it is one of the leading causes of lower respiratory tract infections and hospital visits during infancy and childhood.
Specifications
| RSV Fusion Protein | |
| Monoclonal | |
| 0.1 mg/mL | |
| PBS with 0.1% sodium azide; pH 7.2 | |
| Mouse | |
| Protein A | |
| RUO | |
| Virus | |
| Antibody | |
| IgG2b |
| ELISA, Inhibition Assays, Immunocytochemistry | |
| D74G | |
| Unconjugated | |
| human respiratory syncytial virus; RSV; RSV Fusion protein; Rsvgp08 | |
| Fusion protein of RSV. | |
| 100 μg | |
| Primary | |
| 4°C | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.