missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RSV Fusion Protein Antibody (RSHZ19 (Felvizumab)), DyLight 488, Novus Biologicals™
Shop All Bio Techne ProductsDescription
RSV Fusion Protein Monoclonal specifically detects RSV Fusion Protein in Virus samples. It is validated for ELISA.
Specifications
Specifications
| Antigen | RSV Fusion Protein |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | RSHZ19 (Felvizumab) |
| Conjugate | DyLight 488 |
| Dilution | ELISA |
| Formulation | 50mM Sodium Borate |
| Host Species | Rabbit |
| Immunogen | This antibody was prepared by the humanization of an F protein-specific murine MAb, RSV19. More specifically, molecular techniques were used to insert the complementarity determining regions from murine RSV19 into a human IgG1 heavy and k light chain-variable domain framework. |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?