missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RSV Fusion Protein Antibody (RSHZ19 (Felvizumab)), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-75191CL1
This item is not returnable.
View return policy
Description
RSV Fusion Protein Monoclonal antibody specifically detects RSV Fusion Protein in Virus samples. It is validated for ELISA,Functional Assay
Specifications
| RSV Fusion Protein | |
| Monoclonal | |
| CoraFluor 1 | |
| Human respiratory syncytial virus, RSV, RSV fusion protein, Rsvgp08 | |
| This antibody was prepared by the humanization of an F protein-specific murine MAb, RSV19. More specifically, molecular techniques were used to insert the complementarity determining regions from murine RSV19 into a human IgG1 heavy and k light chain-variable domain framework. | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG κ |
| ELISA, Functional Assay | |
| RSHZ19 (Felvizumab) | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction