missing translation for 'onlineSavingsMsg'
Learn More

RSV Fusion Protein Antibody (RSHZ19 (Felvizumab)), Alexa Fluor™ 750, Novus Biologicals™

Product Code. 18375930 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18375930 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18375930 Supplier Novus Biologicals Supplier No. NBP275190AF750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human Monoclonal Antibody

RSV Fusion Protein Monoclonal specifically detects RSV Fusion Protein in Viral samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RSV Fusion Protein
Applications ELISA
Classification Monoclonal
Clone RSHZ19 (Felvizumab)
Conjugate Alexa Fluor 750
Dilution ELISA
Formulation 50mM Sodium Borate
Host Species Human
Immunogen This antibody was prepared by the humanization of an F protein-specific murine MAb, RSV19. More specifically, molecular techniques were used to insert the complementarity determining regions from murine RSV19 into a human IgG1 heavy and k light chain-variable domain framework.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.