missing translation for 'onlineSavingsMsg'
Learn More

RSV/BRSV Antibody, CoraFluor™ 1, Novus Biologicals™

Product Code. 30150187
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30150187 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30150187 Supplier Novus Biologicals (Bio-Techne) Supplier No. NB10093585CL1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RSV/BRSV Polyclonal antibody specifically detects RSV/BRSV in Virus samples. It is validated for Flow Cytometry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen RSV/BRSV
Applications Flow Cytometry, Western Blot
Classification Polyclonal
Conjugate CoraFluor 1
Formulation PBS
Gene Alias Bovine Respiratory Syncytial Virus BRSV, Bovine RSV
Host Species Rabbit
Immunogen Residues 201-211 [KELLPKVNNHDC] of the 63 kDa BRSV F protein.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C in the dark. Do not freeze.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.