missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RSU1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11207-1-AP-150UL
This item is not returnable.
View return policy
Description
Component of the HBO1 complex which has a histone H4-specific acetyltransferase activity, a reduced activity toward histone H3 and is responsible for the bulk of histone H4 acetylation in vivo. Through chromatin acetylation it may regulate DNA replication and act as a coactivator of TP53-dependent transcription. Specifically represses AR-mediated transcription.Specifications
| RSU1 | |
| Polyclonal | |
| Unconjugated | |
| RSU1 | |
| Ras suppressor protein 1, RSP 1, RSP1, Rsu 1, RSU1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 20163, 6251 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q01730, Q15404 | |
| RSU1 | |
| RSU1 Fusion Protein Ag1693 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction