missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RSRC1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560828
This item is not returnable.
View return policy
Description
Immunogen sequence: KGKDKELHNI KRGESGNIKA GLEHLPPAEQ AKARLQLVLE AAAKADEALK AKERNEEEAK RRKEEDQATL VEQVKRVKEI EAIESD Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 81%.
RSRC1 is a gene that encodes a protein belonging to the serine and arginine rich-related protein family. This protein plays a crucial role in both constitutive and alternative mRNA splicing. Research suggests that this gene may be linked to schizophrenia, and a pseudogene of RSRC1 is present on chromosome 9. Alternative splicing of RSRC1 leads to the formation of multiple transcript variants that encode distinct isoforms. Additionally, RSRC1 has been associated with various disorders, including Autosomal Recessive Non-Syndromic Intellectual Disability and Intellectual Developmental Disorder, Autosomal Recessive 70.
Specifications
| RSRC1 | |
| Polyclonal | |
| Unconjugated | |
| RSRC1 | |
| 1200013F24Rik; arginine and serine rich coiled-coil 1; arginine/serine-rich coiled coil protein 1; arginine/serine-rich coiled-coil 1; arginine/serine-rich coiled-coil protein 1; BM-011; RGD1304968; Rsrc1; Serine/Arginine-related protein 53; SFRS21; splicing factor, arginine/serine-rich 21; Srrp53 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51319 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96IZ7 | |
| RSRC1 | |
| Recombinant protein corresponding to Human RSRC1. Recombinant protein control fragment (Product #RP-100236). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction