missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RSRC1 Polyclonal Antibody

Product Code. 12819042
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12819042 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12819042 Supplier Invitrogen™ Supplier No. PA520899

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is HepG2 cell lysate. PA5-20899 can be used with blocking peptide PEP-1013.

RSRC1 is a gene that encodes a protein belonging to the serine and arginine rich-related protein family. This protein plays a crucial role in both constitutive and alternative mRNA splicing. Research suggests that this gene may be linked to schizophrenia, and a pseudogene of RSRC1 is present on chromosome 9. Alternative splicing of RSRC1 leads to the formation of multiple transcript variants that encode distinct isoforms. Additionally, RSRC1 has been associated with various disorders, including Autosomal Recessive Non-Syndromic Intellectual Disability and Intellectual Developmental Disorder, Autosomal Recessive 70.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RSRC1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene RSRC1
Gene Accession No. Q5PPJ2, Q96IZ7, Q9DBU6
Gene Alias 1200013F24Rik; arginine and serine rich coiled-coil 1; arginine/serine-rich coiled coil protein 1; arginine/serine-rich coiled-coil 1; arginine/serine-rich coiled-coil protein 1; BM-011; RGD1304968; Rsrc1; Serine/Arginine-related protein 53; SFRS21; splicing factor, arginine/serine-rich 21; Srrp53
Gene Symbols RSRC1
Host Species Rabbit
Immunogen A 19 amino acid peptide near the carboxy terminus of human RSRC1.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 361956, 51319, 66880
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.