missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RSPO4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA525615
This item is not returnable.
View return policy
Description
RSPO4 Polyclonal Antibody for Western Blot, IHC (P), Flow
RSPO4 is a member of the R-spondin family of proteins that share a common domain organization consisting of a signal peptide, cysteine-rich/furin-like domain, thrombospondin domain and a C-terminal basic region. The encoded protein may be involved in activation of Wnt/beta-catenin signaling pathways. Mutations in this gene are associated with anonychia congenital. Alternate splicing results in multiple transcript variants.
Specifications
| RSPO4 | |
| Polyclonal | |
| Unconjugated | |
| RSPO4 | |
| A730099F22; A930029K19Rik; C20orf182; CRISTIN4; Cristin-4; Cysteine-rich and single thrombospondin domain-containing protein 4; hRspo4; mCristin-4; roof plate-specific spondin-4; RSPO4; R-spondin 4; R-spondin family, member 4; R-spondin-4; R-spondin-like protein | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 343637 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q2I0M5 | |
| RSPO4 | |
| KLH conjugated synthetic peptide between 199-228 amino acids from the C-terminal region of human RSPO4. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction