missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RSPO4 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA547579
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
RSPO4 is a member of the R-spondin family of proteins that share a common domain organization consisting of a signal peptide, cysteine-rich/furin-like domain, thrombospondin domain and a C-terminal basic region. The encoded protein may be involved in activation of Wnt/beta-catenin signaling pathways. Mutations in this gene are associated with anonychia congenital. Alternate splicing results in multiple transcript variants.
Specifications
| RSPO4 | |
| Polyclonal | |
| Unconjugated | |
| RSPO4 | |
| A730099F22; A930029K19Rik; C20orf182; CRISTIN4; Cristin-4; Cysteine-rich and single thrombospondin domain-containing protein 4; hRspo4; mCristin-4; roof plate-specific spondin-4; RSPO4; R-spondin 4; R-spondin family, member 4; R-spondin-4; R-spondin-like protein | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 228770 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Neutralization | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q8BJ73 | |
| RSPO4 | |
| E. coli-derived recombinant mouse R-Spondin 4 Tyr21-Pro197. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction