missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RSPO4 Monoclonal Antibody (435203)
GREENER_CHOICE

Rat Monoclonal Antibody

Brand:  Invitrogen™ MA524160

Product Code. 15533852

  • £442.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

In Western blots, 50-100% cross-reactivity with recombinant human (rh) R-Spondin 4 is observed and no cross-reactivity with recombinant mouse (rm) R-Spondin 1, rmR-Spondin 3, or rhR-Spondin 2 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.

RSPO4 is a member of the R-spondin family of proteins that share a common domain organization consisting of a signal peptide, cysteine-rich/furin-like domain, thrombospondin domain and a C-terminal basic region. The encoded protein may be involved in activation of Wnt/beta-catenin signaling pathways. Mutations in this gene are associated with anonychia congenital. Alternate splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RSPO4
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
Q8BJ73
RSPO4
E. coli-derived recombinant mouse R-Spondin 4 Tyr21-Pro197.
100 μg
Primary
Mouse
Antibody
IgG2b
Western Blot
435203
Unconjugated
RSPO4
A730099F22; A930029K19Rik; C20orf182; CRISTIN4; Cristin-4; Cysteine-rich and single thrombospondin domain-containing protein 4; hRspo4; mCristin-4; roof plate-specific spondin-4; RSPO4; R-spondin 4; R-spondin family, member 4; R-spondin-4; R-spondin-like protein
Rat
Protein A/G
RUO
228770
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.