missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RSPO2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17781-1-AP-150UL
This item is not returnable.
View return policy
Description
R-spondins signaling.Specifications
| RSPO2 | |
| Polyclonal | |
| Unconjugated | |
| RSPO2 | |
| CRISTIN2, hRspo2, R spondin 2, Roof plate specific spondin 2, RSPO2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 239405, 340419, 500863 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.39 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6UXX9, Q8BFU0 | |
| RSPO2 | |
| RSPO2 Fusion Protein Ag11216 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction