missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RSPH9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA545967
This item is not returnable.
View return policy
Description
Peptide sequence: VKGRFMGDPS YEYEHTELQK VNEGEKVFEE EIVVQIKEET RLVSVIDQID Sequence homology: Cow: 100%; Dog: 86%; Horse: 86%; Human: 100%; Mouse: 93%; Pig: 100%; Rabbit: 79%; Rat: 79%.
This gene encodes a protein thought to be a component of the radial spoke head in motile cilia and flagella. Mutations in this gene are associated with primary ciliary dyskinesia 12. Alternative splicing results in multiple transcript variants.
Specifications
| RSPH9 | |
| Polyclonal | |
| Unconjugated | |
| RSPH9 | |
| 1700027N10Rik; C23H6orf206; C6orf206; CILD12; MRPS18AL1; radial spoke head 9 homolog; radial spoke head 9 homolog (Chlamydomonas); radial spoke head component 9; radial spoke head protein 9 homolog; RGD1310693; RSPH9; similar to CG31803-PA | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 221421 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9H1X1 | |
| RSPH9 | |
| Synthetic peptide directed towards the middle region of human RSPH9. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction