missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RSPH4A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA546058
This item is not returnable.
View return policy
Description
Peptide sequence: APPQSDRTTS VIPEAGTPYP DPLEQSSDKR ESTPHHTSQS EGNTFQQSQQ Sequence homology: Human: 100%.
This gene encodes a protein that appears to be a component the radial spoke head, as determined by homology to similar proteins in the biflagellate alga Chlamydomonas reinhardtii and other ciliates. Radial spokes, which are regularly spaced along cilia, sperm, and flagella axonemes, consist of a thin 'stalk' and a bulbous 'head' that form a signal transduction scaffold between the central pair of microtubules and dynein. Mutations in this gene cause primary ciliary dyskinesia 1, a disease arising from dysmotility of motile cilia and sperm. Alternative splicing results in multiple transcript variants.
Specifications
| RSPH4A | |
| Polyclonal | |
| Unconjugated | |
| RSPH4A | |
| A230081C05; CILD11; dJ412I7.1; radial spoke head 4 homolog A; radial spoke head 4 homolog A (Chlamydomonas); radial spoke head protein 4 homolog A; Radial spoke head-like protein 3; radial spokehead-like 3; RSHL3; Rsph4a; RSPH6B | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 345895 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q5TD94 | |
| RSPH4A | |
| Synthetic peptide directed towards the N-terminal of human RSPH4A (aa 106-155). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction