missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RSPH4A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA556666
This item is not returnable.
View return policy
Description
Immunogen sequence: RPWEGKTAAS PQYSEPESSE PLEAKQGPET GRQSRSSRPW SPQSRAKTPL GGPAGPETSS PAPVSPREPS SSP Highest antigen sequence identity to the following orthologs: Mouse - 41%, Rat - 37%.
This gene encodes a protein that appears to be a component the radial spoke head, as determined by homology to similar proteins in the biflagellate alga Chlamydomonas reinhardtii and other ciliates. Radial spokes, which are regularly spaced along cilia, sperm, and flagella axonemes, consist of a thin 'stalk' and a bulbous 'head' that form a signal transduction scaffold between the central pair of microtubules and dynein. Mutations in this gene cause primary ciliary dyskinesia 1, a disease arising from dysmotility of motile cilia and sperm. Alternative splicing results in multiple transcript variants.
Specifications
| RSPH4A | |
| Polyclonal | |
| Unconjugated | |
| RSPH4A | |
| A230081C05; CILD11; dJ412I7.1; radial spoke head 4 homolog A; radial spoke head 4 homolog A (Chlamydomonas); radial spoke head protein 4 homolog A; Radial spoke head-like protein 3; radial spokehead-like 3; RSHL3; Rsph4a; RSPH6B | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 345895 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5TD94 | |
| RSPH4A | |
| Recombinant protein corresponding to Human RSPH4A. Recombinant protein control fragment (Product #RP-94636). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction