missing translation for 'onlineSavingsMsg'
Learn More

RSL3, MedChemExpress

Product Code. 16492848
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
2mg
50mg
5mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16492848 10 mg 10mg
16459506 50 mg 50mg
16422546 25 mg 25mg
16420006 2 mg 2mg
16415597 5 mg 5mg
16414266 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16492848 Supplier Medchem Express Supplier No. HY100218A
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress RSL3 ((1S,3R)-RSL3) is an inhibitor of glutathione peroxidase 4 (GPX4) (ferroptosis activator), reduces the expression of GPX4 protein, and induces ferroptotic death of head and neck cancer cell. RSL3 increases the expression of p62 and Nrf2 and inactivates Keap1 in HN3-rslR cells.

RSL3 ((1S,3R)-RSL3) is an inhibitor of glutathione peroxidase 4 (GPX4) (ferroptosis activator), reduces the expression of GPX4 protein, and induces ferroptotic death of head and neck cancer cell. RSL3 increases the expression of p62 and Nrf2 and inactivates Keap1 in HN3-rslR cells.

Specifications

Chemical Name or Material RSL3
CAS 1219810-16-8
Color White
Purity Grade Notes Research
Molecular Formula C23H21ClN2O5
Quantity 10 mg
Synonym (1S,3R)-RSL3
Solubility Information DMSO : 250 mg/mL (567.05 mM; Need ultrasonic)
SMILES O=C([C@H]1CC2=C([C@H](C3=CC=C(C(OC)=O)C=C3)N1C(CCl)=O)NC4=C2C=CC=C4)OC
Molecular Weight (g/mol) 440.88
Formula Weight 440.88
Percent Purity 98.1%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.