missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RSK4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA582123
This item is not returnable.
View return policy
Description
Immunogen sequence: YTMLAGYTPF ANGPNDTPEE ILLRIGNGKF SLSGGNWDNI SDGAKDLLSH MLHMDPHQRY TAEQILKHSW ITHRDQLPND QPKRNDVSHV VKGAMVATYS ALTHKTF Highest antigen sequence indentity to the following orthologs: Rat - 75%, Mouse - 72%.
RPS6KA6 (RSK4) is a member of the RSK kinases family. RSK family members are activated by ERKs and mitogens and are unusual in that they contain 2 non-identical kinase domains. RSK4 encodes a member of ribosomal S6 kinase family, serine-threonine protein kinases which are regulated by growth factors. The encoded protein may be distinct from other members of this family, however, as studies suggest it is not growth factor dependent and may not participate in the same signaling pathways.
Specifications
| RSK4 | |
| Polyclonal | |
| Unconjugated | |
| Rps6ka6 | |
| 2610524K04Rik; 90 kDa ribosomal protein S6 kinase 6; p90-RSK 6; p90RSK6; pp90RSK4; RGD1560817; ribosomal protein S6 kinase A6; ribosomal protein S6 kinase alpha-6; ribosomal protein S6 kinase polypeptide 6; ribosomal protein S6 kinase, 90kDa, polypeptide 6; ribosomal S6 kinase 4; Rps6ka6; RSK4; RSK-4; S6KA6; S6K-alpha 6; S6K-alpha-6 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 27330 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UK32 | |
| Rps6ka6 | |
| Recombinant protein corresponding to Human RPS6KA6. Recombinant protein control fragment (Product #RP-92328). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction