missing translation for 'onlineSavingsMsg'
Learn More

RSK4 Antibody (6F2), Novus Biologicals™

Product Code. 18328509 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18328509 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18328509 Supplier Novus Biologicals Supplier No. H00027330M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

RSK4 Monoclonal antibody specifically detects RSK4 in Human, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen RSK4
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 6F2
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_055311.1
Gene Alias EC 2.7.11, EC 2.7.11.1, p90-RSK 6, p90RSK6, pp90RSK4, PP90RSK4,90 kDa ribosomal protein S6 kinase 6, ribosomal protein S6 kinase alpha-6, ribosomal protein S6 kinase, 90kDa, polypeptide 6, Ribosomal S6 kinase 4, RSK-4, RSK4ribosomal protein S6 kinase, 90kD, polypeptide 6, S6K-alpha 6, S6K-alpha-6
Host Species Mouse
Immunogen RPS6KA6 (NP_055311.1, 636 a.a. ~ 745 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. RIGNGKFSLSGGNWDNISDGAKDLLSHMLHMDPHQRYTAEQILKHSWITHRDQLPNDQPKRNDVSHVVKGAMVATYSALTHKTFQPVLEPVAASSLAQRRSMKKRTSTGL
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 27330
Target Species Human, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.