missing translation for 'onlineSavingsMsg'
Learn More

RSK3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16883084 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16883084 150 μL 150µL
16893084 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16883084 Supplier Proteintech Supplier No. 144461AP150UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This gene encodes a member of the RSK (ribosomal S6 kinase) family of serine/threonine kinases. This kinase contains 2 nonidentical kinase catalytic domains and phosphorylates various substrates, including members of the mitogen-activated kinase (MAPK) signalling pathway. The activity of this protein has been implicated in controlling cell growth and differentiation. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.

Specifications

Antigen RSK3
Applications Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene RPS6KA2
Gene Accession No. Q15349, Q9WUT3
Gene Alias HU 2, MAPKAP kinase 1c, MAPKAPK 1c, MAPKAPK1C, p90 RSK 2, p90 RSK3, p90RSK2, pp90RSK3, Ribosomal S6 kinase 3, RPS6KA2, RSK, RSK 3, RSK3, S6K alpha, S6K alpha 2, S6K alpha2
Gene Symbols RPS6KA2
Host Species Rabbit
Immunogen RSK3 Fusion Protein Ag5862
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117269, 20112, 6196
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.