missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
This gene encodes a member of the RSK (ribosomal S6 kinase) family of serine/threonine kinases. This kinase contains 2 nonidentical kinase catalytic domains and phosphorylates various substrates, including members of the mitogen-activated kinase (MAPK) signalling pathway. The activity of this protein has been implicated in controlling cell growth and differentiation. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
Specifications
Specifications
| Antigen | RSK3 |
| Applications | Immunofluorescence, Immunoprecipitation, Immunocytochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.25 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | RPS6KA2 |
| Gene Accession No. | Q15349, Q9WUT3 |
| Gene Alias | HU 2, MAPKAP kinase 1c, MAPKAPK 1c, MAPKAPK1C, p90 RSK 2, p90 RSK3, p90RSK2, pp90RSK3, Ribosomal S6 kinase 3, RPS6KA2, RSK, RSK 3, RSK3, S6K alpha, S6K alpha 2, S6K alpha2 |
| Gene Symbols | RPS6KA2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?