missing translation for 'onlineSavingsMsg'
Learn More

RSK3 Antibody, DyLight 755, Novus Biologicals™

Product Code. 18336186 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18336186 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18336186 Supplier Novus Biologicals Supplier No. NBP298853IR

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RSK3 Polyclonal specifically detects RSK3 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RSK3
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 755
Formulation 50 mM Sodium Borate
Gene Alias EC 2.7.11, EC 2.7.11.1, HU-2, MAP kinase-activated protein kinase 1c, MAPK-activated protein kinase 1c, MAPKAP kinase 1c, MAPKAPK-1c, MAPKAPK1C, p90-RSK 2, p90RSK2, p90-RSK3, ribosomal protein S6 kinase alpha 2, ribosomal protein S6 kinase alpha-2, ribosomal protein S6 kinase, 90kD, polypeptide 2, ribosomal protein S6 kinase, 90kDa, polypeptide 2, Ribosomal S6 kinase 3,90 kDa ribosomal protein S6 kinase 2, RSK, RSK-3, RSK3pp90RSK3, S6K-alpha, S6K-alpha2, S6K-alpha-2
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human RSK3.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 6196
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.